Ethyl-4-amino-2-mercaptopyrimidine-5-carboxylate

Lieferant: Apollo Scientific

OR21285-5G
APOSOR21285-5GEA 60.9 EUR
APOSOR21285-5G
Ethyl-4-amino-2-mercaptopyrimidine-5-carboxylate
Ethyl-4-amino-2-mercaptopyrimidine-5-carboxylate
Formel: C₇H₉N₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00023227
CAS-Nummer: 774-07-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR