2,6-Dibrom-4-cyclohexylanilin

Lieferant: Apollo Scientific

OR21303-1G
APOSOR21303-1GEA 87 EUR
APOSOR21303-1G
2,6-Dibrom-4-cyclohexylanilin
2,6-Dibrom-4-cyclohexylanilin
Formel: C₁₂H₁₅Br₂N
Lagertemperatur: Raumtemperatur
CAS-Nummer: 175135-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR