Methyl-3-hydroxy-2-benzo[b]thiophencarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
OR21316-1G
APOSOR21316-1GEA
109
EUR
APOSOR21316-1G
Methyl-3-hydroxy-2-benzo[b]thiophencarboxylat
Methyl-3-hydroxy-2-benzo[b]thiophencarboxylat
Formel:
C₁₀H₈O₃S Schmelzpunkt: 107…108 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00067732 CAS-Nummer: 13134-76-4 EINECS: 236-066-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...