3-Amino-4-brom-5-methylisoxazol

Lieferant: Apollo Scientific

Synonyme: 4-bromo-5-methylisoxazol-3-amine

OR21353-1G
APOSOR21353-1GEA 50.9 EUR
APOSOR21353-1G
3-Amino-4-brom-5-methylisoxazol
3-Amino-4-bromo-5-methylisoxazole
Formel: C₄H₅BrN₂O
Molecular Weight: 177,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052553
CAS-Nummer: 5819-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR