2-(Hydroxymethyl)-6-methylpyridin-3-ol 95+%
Lieferant: Apollo Scientific
Synonyme:
2,6-Lutidine-α2,3-diol
OR21354-5G
APOSOR21354-5GEA
85.7
EUR
APOSOR21354-5G
2-(Hydroxymethyl)-6-methylpyridin-3-ol 95+%
2-(Hydroxymethyl)-6-methylpyridin-3-ol
Formel:
C₇H₉NO₂ Molecular Weight: 139,154 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006350 CAS-Nummer: 42097-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...