3-(4-Bromothien-2-yl)-1-(4-chlorophenyl)prop-2-en-1-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-2-[3-(4-chlorophenyl)-3-oxoprop-1-en-1-yl]thiophene, 1-[(4-Bromothien-2-yl)acryloyl]-4-chlorobenzene
OR21376-250MG
APOSOR21376-250MGEA
116
EUR
APOSOR21376-250MG
3-(4-Bromothien-2-yl)-1-(4-chlorophenyl)prop-2-en-1-one
3-(4-Bromothien-2-yl)-1-(4-chlorophenyl)prop-2-en-1-one
Formel:
C₁₃H₈BrClOS Lagertemperatur: Kühlschrank |
MDL:
MFCD00117349 CAS-Nummer: 219689-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...