N'-Hydroxy-N-(5-methylisoxazol-3-yl)formamidine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-{[(Hydroxyimino)methyl]amino}-5-methylisoxazole, N'-Hydroxy-N-(5-methyl-1,2-oxazol-3-yl)imidoformamide

OR21386-250MG
APOSOR21386-250MGEA 97.3 EUR
APOSOR21386-250MG
N'-Hydroxy-N-(5-methylisoxazol-3-yl)formamidine
N'-Hydroxy-N-(5-methylisoxazol-3-yl)formamidine
Formel: C₅H₇N₃O₂
Molecular Weight: 141,128 g/mol
Schmelzpunkt: 185…187 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00105523
CAS-Nummer: 382136-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR