8-Chloronaphthalene-1-sulphonic acid 97%

Lieferant: Apollo Scientific
Gefahr

OR21393-250MG
APOSOR21393-250MGEA 40.6 EUR
APOSOR21393-250MG
8-Chloronaphthalene-1-sulphonic acid 97%
8-Chloronaphthalene-1-sulphonic acid
Formel: C₁₀H₇ClO₃S
Molecular Weight: 242,679 g/mol
Schmelzpunkt: 145…147 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052379
CAS-Nummer: 145-74-4
EINECS: 205-661-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR