6-Amino-2-methylchinolin

Lieferant: Apollo Scientific

Synonyme: 2-methylquinolin-6-amine

OR21398-1G
APOSOR21398-1GEA 62.5 EUR
APOSOR21398-1G
6-Amino-2-methylchinolin
6-Amino-2-methylquinoline
Formel: C₁₀H₁₀N₂
Molecular Weight: 158,20 g/mol
Schmelzpunkt: 189…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052600
CAS-Nummer: 65079-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR