2-Chloro-N-(4-iodophenyl)acetamide

Lieferant: Apollo Scientific

Synonyme: 4'-Iodo-2-chloroacetoanilide, N1-(4-Iodophenyl)-2-chloroacetamide, Acetamide,2-chloro-N-(4-iodophenyl)-

OR21404-1G
APOSOR21404-1GEA 101 EUR
APOSOR21404-1G
2-Chloro-N-(4-iodophenyl)acetamide
2-Chloro-N-(4-iodophenyl)acetamide
Formel: C₈H₇ClINO
Molecular Weight: 295,51 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00171531
CAS-Nummer: 2564-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR