2-Chlor-4'-nitroacetanilid

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(Chloroacetyl)-4-nitroaniline

OR21441-1G
APOSOR21441-1GEA 21.9 EUR
APOSOR21441-1G
2-Chlor-4'-nitroacetanilid
2-Chlor-4'-nitroacetanilid
Formel: C₈H₇ClN₂O₃
Molecular Weight: 214,61 g/mol
Schmelzpunkt: 185…186 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00031337
CAS-Nummer: 17329-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR