2-Chloro-N-(4-chloro-2-methylphenyl)acetamide
Lieferant: Apollo Scientific
OR21445-1G
APOSOR21445-1GEA
212
EUR
APOSOR21445-1G
2-Chloro-N-(4-chloro-2-methylphenyl)acetamide
2-Chloro-N-(4-chloro-2-methylphenyl)acetamide
Formel:
C₉H₉Cl₂NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD00018906 CAS-Nummer: 62593-77-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...