2-Cyano-N-methyl-acetamide
Lieferant: Apollo Scientific
|
Achtung
|
OR21454-10G
OR21454-1G
OR21454-5G
APOSOR21454-10GEA
0
EUR
APOSOR21454-10G
APOSOR21454-1G
APOSOR21454-5G
2-Cyano-N-methyl-acetamide
2-Cyano-N-methyl-acetamide
Formel:
C₄H₆N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00090119 CAS-Nummer: 6330-25-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...