5-Bromo-3,4-dimethylbenzene-1,2-diamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-3,4-dimethylphenylene-1,2-diamine, 6-Bromo-3,4-diamino-o-xylene
OR21478-250MG
APOSOR21478-250MGEA
21.9
EUR
APOSOR21478-250MG
5-Bromo-3,4-dimethylbenzene-1,2-diamine
5-Bromo-3,4-dimethylbenzene-1,2-diamine
Formel:
C₈H₁₁BrN₂ Molecular Weight: 215,09 g/mol Schmelzpunkt: 87…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052696 CAS-Nummer: 107100-16-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...