3-(2-Aminothien-5-yl)pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-(pyridin-3-yl)thiophene, 5-(Pyridin-3-yl)thien-2-amine

OR2148-250MG
APOSOR2148-250MGEA 980 EUR
APOSOR2148-250MG
3-(2-Aminothien-5-yl)pyridine
3-(2-Aminothien-5-yl)pyridine
Formel: C₉H₈N₂S
Molecular Weight: 176,238 g/mol
Schmelzpunkt: 126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09038085
CAS-Nummer: 837376-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR