(6-Chloroimidazo[1,2-b]pyridazin-3-yl)(phenyl)methanone

Lieferant: Apollo Scientific
Achtung

OR21487-1G OR21487-50MG OR21487-250MG OR21487-500MG
APOSOR21487-1GEA 314 EUR
APOSOR21487-1G APOSOR21487-50MG APOSOR21487-250MG APOSOR21487-500MG
(6-Chloroimidazo[1,2-b]pyridazin-3-yl)(phenyl)methanone
(6-Chloroimidazo[1,2-b]pyridazin-3-yl)(phenyl)methanone
Formel: C₁₃H₈ClN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00108116
CAS-Nummer: 90734-72-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR