4,5-Dimethylpyridazine-3,6-diol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,6-Dihydroxy-4,5-dimethyl-1,2-diazine, 3,6-Dihydroxy-4,5-dimethylpyridazine
OR21499-250MG
APOSOR21499-250MGEA
21.9
EUR
APOSOR21499-250MG
4,5-Dimethylpyridazine-3,6-diol
4,5-Dimethylpyridazine-3,6-diol
Formel:
C₆H₈N₂O₂ Molecular Weight: 140,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067744 CAS-Nummer: 5754-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...