(2-Brom-5-chlorphenyl)essigsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2-Bromo-5-chlorophenyl)acetic acid
OR2153-100G
OR2153-10G
OR2153-25G
APOSOR2153-100GEA
545
EUR
APOSOR2153-100G
APOSOR2153-10G
APOSOR2153-25G
(2-Brom-5-chlorphenyl)essigsäure
(2-Brom-5-chlorphenyl)essigsäure
Formel:
C₈H₆BrClO₂ Schmelzpunkt: 101…103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153890 CAS-Nummer: 81682-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...