O411 98+%

Lieferant: Apollo Scientific

Synonyme: 4-[(4-Methoxyphenyl)methoxy]benzeneacetonitrile, 2-(4-((4-Methoxybenzyl)oxy)phenyl)acetonitrile, O4I1

OR21534-250MG
APOSOR21534-250MGEA 223 EUR
APOSOR21534-250MG
O411 98+%
O411
Formel: C₁₆H₁₅NO₂
Molecular Weight: 253,30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00067778
CAS-Nummer: 175135-47-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR