2-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridin-3-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2,5-Dimethyl-1H-pyrrol-1-yl)-3-hydroxypyridine, 2,5-Dimethyl-1-(3-hydroxypyridin-2-yl)-1H-pyrrole

OR21535-1G
APOSOR21535-1GEA 174 EUR
APOSOR21535-1G
2-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridin-3-ol
2-(2,5-Dimethyl-1H-pyrrol-1-yl)pyridin-3-ol
Formel: C₁₁H₁₂N₂O
Molecular Weight: 188.226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00067779
CAS-Nummer: 175135-48-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR