2-Chloro-3-[(5-nitro-2-pyridyl)oxy]pyridine

Lieferant: Apollo Scientific

OR21555-1G
APOSOR21555-1GEA 198 EUR
APOSOR21555-1G
2-Chloro-3-[(5-nitro-2-pyridyl)oxy]pyridine
2-Chloro-3-[(5-nitro-2-pyridyl)oxy]pyridine
Formel: C₁₀H₆ClN₃O₃
Molecular Weight: 251,626 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00067785
CAS-Nummer: 219928-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR