2-Brom-4-chlorphenol 97%

Lieferant: Apollo Scientific
Achtung

OR2160-500G
APOSOR2160-500GEA 253 EUR
APOSOR2160-500G
2-Brom-4-chlorphenol 97%
2-Brom-4-chlorphenol
Formel: C₆H₄BrClO
Lagertemperatur: Raumtemperatur
MDL: MFCD00002319
CAS-Nummer: 695-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR