2-[4-(tert-Butyl)phenyl]-5-(2-chloro-5-nitrophenyl)-1,3,4-oxadiazole

Lieferant: Apollo Scientific
Achtung

OR21617-100MG
APOSOR21617-100MGEA 21.9 EUR
APOSOR21617-100MG
2-[4-(tert-Butyl)phenyl]-5-(2-chloro-5-nitrophenyl)-1,3,4-oxadiazole
2-[4-(tert-Butyl)phenyl]-5-(2-chloro-5-nitrophenyl)-1,3,4-oxadiazole
Formel: C₁₈H₁₆ClN₃O₃
Molecular Weight: 357,791 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00117794
CAS-Nummer: 243644-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR