2-(4-Chlorphenylthio)nicotinsäure

Lieferant: Apollo Scientific

OR21702-1G
APOSOR21702-1GEA 65.4 EUR
APOSOR21702-1G
2-(4-Chlorphenylthio)nicotinsäure
2-(4-Chlorphenylthio)nicotinsäure
Formel: C₁₂H₈ClNO₂S
Schmelzpunkt: 219…221 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052117
CAS-Nummer: 955-54-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR