2-[(4-Chlorophenyl)thio]pyridine-3-carbonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR21705-1G
APOSOR21705-1GEA
0
EUR
APOSOR21705-1G
2-[(4-Chlorophenyl)thio]pyridine-3-carbonyl chloride
2-[(4-Chlorophenyl)thio]pyridine-3-carbonyl chloride
Formel:
C₁₂H₇Cl₂NOS Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067836 CAS-Nummer: 97936-44-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...