[(8-Chlor-1-naphthyl)sulfanyl]essigsäure
Lieferant: Apollo Scientific
OR21716-1G
APOSOR21716-1GEA
62.5
EUR
APOSOR21716-1G
[(8-Chlor-1-naphthyl)sulfanyl]essigsäure
(8-Chloro-1-naphthylthio)acetic acid
Formel:
C₁₂H₉ClO₂S Molecular Weight: 252.71 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043901 CAS-Nummer: 129-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...