2-Bromo-3'-chloro-4'-methylpropiophenone, tech

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-(3-chloro-4-methylphenyl)propan-1-one

OR21756-1G
APOSOR21756-1GEA 0 EUR
APOSOR21756-1G
2-Bromo-3'-chloro-4'-methylpropiophenone, tech
2-Bromo-3'-chloro-4'-methylpropiophenone
Formel: C₁₀H₁₀BrClO
Molecular Weight: 261.543 g/mol
Siedepunkt: 121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067862
CAS-Nummer: 175135-93-0
UN: 3261
ADR: 8 I

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR