Dimethyl biphenyl-4,4'-dicarboxylate 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: Dimethyl [1,1'-biphenyl]-4,4'-dicarboxylate, 4,4'-Biphenyldicarboxylic Acid Dimethyl Ester, 4,4'-Bibenzoic Acid Dimethyl Ester, Dimethyl-4,4'-biphenyldicarboxylate

OR21762-25G OR21762-100G
APOSOR21762-25GEA 56.7 EUR
APOSOR21762-25G APOSOR21762-100G
Dimethyl biphenyl-4,4'-dicarboxylate 99%
Dimethyl biphenyl-4,4'-dicarboxylate
Formel: C₁₆H₁₄O₄
Molecular Weight: 270.28 g/mol
Schmelzpunkt: 213…215 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017201
CAS-Nummer: 792-74-5
EINECS: 212-341-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR