Methyl-(3-methoxyphenyl)acetat 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-Methoxyphenylacetate
OR21784-1G
OR21784-25G
OR21784-5G
APOSOR21784-1GEA
21.9
EUR
APOSOR21784-1G
APOSOR21784-25G
APOSOR21784-5G
Methyl-(3-methoxyphenyl)acetat 98%
Methyl-(3-methoxyphenyl)acetat
Formel:
C₁₀H₁₂O₃ Siedepunkt: 86…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017205 CAS-Nummer: 18927-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...