2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2,3-dihydro-1H-pyrrol-3-yl]acetic acid

Lieferant: Apollo Scientific

OR21810-1G
APOSOR21810-1GEA 183 EUR
APOSOR21810-1G
2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2,3-dihydro-1H-pyrrol-3-yl]acetic acid
2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2,3-dihydro-1H-pyrrol-3-yl]acetic acid
Formel: C₉H₁₃NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00173859
CAS-Nummer: 77978-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR