4-Methoxy-7-methylindane-5-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR21831-1G
APOSOR21831-1GEA 72.5 EUR
APOSOR21831-1G
4-Methoxy-7-methylindane-5-carbonitrile
4-Methoxy-7-methylindane-5-carbonitrile
Formel: C₁₂H₁₃NO
Molecular Weight: 187,238 g/mol
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173681
CAS-Nummer: 175136-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR