7-Methyl-4-indanyl acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Methyl-2,3-dihydro-1H-inden-4-yl acetate

OR21833-1G
APOSOR21833-1GEA 87 EUR
APOSOR21833-1G
7-Methyl-4-indanyl acetate
7-Methyl-4-indanyl acetate
Formel: C₁₂H₁₄O₂
Molecular Weight: 190,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173682
CAS-Nummer: 175136-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR