1-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}-1-ethanone

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-[(3,4-Dichlorobenzyl)oxy]acetophenone

OR21843-250MG
APOSOR21843-250MGEA 21.9 EUR
APOSOR21843-250MG
1-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}-1-ethanone
1-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}-1-ethanone
Formel: C₁₅H₁₂Cl₂O₂
Molecular Weight: 295,17 g/mol
Schmelzpunkt: 83…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173669
CAS-Nummer: 170916-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR