4-[(3,4-Dichlorbenzyl)oxy]benzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-((3,4-Dichlorobenzyl)oxy)benzaldehyde
OR21848-1G
APOSOR21848-1GEA
33.5
EUR
APOSOR21848-1G
4-[(3,4-Dichlorbenzyl)oxy]benzaldehyd
4-[(3,4-Dichlorbenzyl)oxy]benzaldehyd
Formel:
C₁₄H₁₀Cl₂O₂ Molecular Weight: 281,1340 g/mol Schmelzpunkt: 92…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173652 CAS-Nummer: 66742-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...