3-Amino-2-(2,3-dimethylphenoxy)pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2,3-Dimethylphenoxy)pyridin-3-amine

OR21861-250MG
APOSOR21861-250MGEA 84.1 EUR
APOSOR21861-250MG
3-Amino-2-(2,3-dimethylphenoxy)pyridine
3-Amino-2-(2,3-dimethylphenoxy)pyridine
Formel: C₁₃H₁₄N₂O
Molecular Weight: 214,263 g/mol
Schmelzpunkt: 143…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173657
CAS-Nummer: 175136-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR