5-Methylnicotinsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Methyl-3-pyridincarbonsäure, 5-Methylpyridin-3-carbonsäure

OR21864-1G OR21864-25G OR21864-5G
APOSOR21864-1GEA 21.9 EUR
APOSOR21864-1G APOSOR21864-25G APOSOR21864-5G
5-Methylnicotinsäure 97%
5-Methylnicotinsäure
Formel: C₇H₇NO₂
Molecular Weight: 137.14 g/mol
Schmelzpunkt: 210…212 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00829036
CAS-Nummer: 3222-49-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR