3-Amino-2-nitropyridin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Nitropyridin-3-amin, 2-Nitro-3-pyridylamin, 2-Nitropyridin-3-ylamin

OR21871-100G
APOSOR21871-100GEA 192 EUR
APOSOR21871-100G
3-Amino-2-nitropyridin 98%
3-Amino-2-nitropyridin
Formel: C₅H₅N₃O₂
Molecular Weight: 139.11 g/mol
Schmelzpunkt: 191…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00044102
CAS-Nummer: 13269-19-7
EINECS: 236-260-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR