4-Hydroxy-3-nitropyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Nitro-4-pyridinol
OR21872-500G
OR21872-100G
OR21872-25G
OR21872-1KG
OR21872-5KG
APOSOR21872-500GEA
367
EUR
APOSOR21872-500G
APOSOR21872-100G
APOSOR21872-25G
APOSOR21872-1KG
APOSOR21872-5KG
4-Hydroxy-3-nitropyridin
4-Hydroxy-3-nitropyridine
Formel:
C₅H₄N₂O₃ Molecular Weight: 140.10 g/mol Schmelzpunkt: 280…285 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128874 CAS-Nummer: 5435-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...