4-Hydroxy-3-nitropyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Nitro-4-pyridinol

OR21872-500G OR21872-100G OR21872-25G OR21872-1KG OR21872-5KG
APOSOR21872-500GEA 367 EUR
APOSOR21872-500G APOSOR21872-100G APOSOR21872-25G APOSOR21872-1KG APOSOR21872-5KG
4-Hydroxy-3-nitropyridin
4-Hydroxy-3-nitropyridine
Formel: C₅H₄N₂O₃
Molecular Weight: 140.10 g/mol
Schmelzpunkt: 280…285 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00128874
CAS-Nummer: 5435-54-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR