N'-Hydroxypyridine-4-carboximidamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Hydroxyisonicotinimidamide, 4-Pyridylamidoxime, Pyridine-4-amidoxime, N'-Hydroxy-4-pyridinecarboximidamide
OR21874-1G
APOSOR21874-1GEA
21.9
EUR
APOSOR21874-1G
N'-Hydroxypyridine-4-carboximidamide
N'-Hydroxypyridine-4-carboximidamide
Formel:
C₆H₇N₃O Molecular Weight: 137,142 g/mol Schmelzpunkt: 193…195 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00125873 CAS-Nummer: 1594-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...