5-Bromo-2-methyl-4-nitro-1H-imidazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-methyl-4-nitroimidazole

OR21890-1G
APOSOR21890-1GEA 21.9 EUR
APOSOR21890-1G
5-Bromo-2-methyl-4-nitro-1H-imidazole
5-Bromo-2-methyl-4-nitro-1H-imidazole
Formel: C₄H₄BrN₃O₂
Molecular Weight: 205,997 g/mol
Schmelzpunkt: 256…257 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00156130
CAS-Nummer: 18874-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR