2-Heptylcyclopentan-1-one oxime

Lieferant: Apollo Scientific

OR21896-1G OR21896-5G OR21896-10G OR21896-25G OR21896-50G
APOSOR21896-1GEA 0 EUR
APOSOR21896-1G APOSOR21896-5G APOSOR21896-10G APOSOR21896-25G APOSOR21896-50G
2-Heptylcyclopentan-1-one oxime
2-Heptylcyclopentan-1-one oxime
Formel: C₁₂H₂₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09998437
CAS-Nummer: 165385-93-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR