6-Chloro-3-pyridineacetic acid 95%

Lieferant: Apollo Scientific
Achtung

OR2190-5G
APOSOR2190-5GEA 23.2 EUR
APOSOR2190-5G
6-Chloro-3-pyridineacetic acid 95%
6-Chloro-3-pyridineacetic acid
Formel: C₇H₆ClNO₂
Schmelzpunkt: 169…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863172
CAS-Nummer: 39891-13-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR