4-Propylcyclohexanol (cis/trans-Gemisch)

Lieferant: Apollo Scientific

OR21917-1G OR21917-250MG OR21917-5G OR21917-10G OR21917-25G
APOSOR21917-1GEA 74.1 EUR
APOSOR21917-1G APOSOR21917-250MG APOSOR21917-5G APOSOR21917-10G APOSOR21917-25G
4-Propylcyclohexanol (cis/trans-Gemisch)
4-Propylcyclohexanol (cis- and trans- mixture)
Formel: C₉H₁₈O
Lagertemperatur: Raumtemperatur
MDL: MFCD00091618
CAS-Nummer: 52204-65-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR