4-Propylcyclohexanol (cis/trans-Gemisch)
Lieferant: Apollo Scientific
OR21917-1G
OR21917-250MG
OR21917-5G
OR21917-10G
OR21917-25G
APOSOR21917-1GEA
74.1
EUR
APOSOR21917-1G
APOSOR21917-250MG
APOSOR21917-5G
APOSOR21917-10G
APOSOR21917-25G
4-Propylcyclohexanol (cis/trans-Gemisch)
4-Propylcyclohexanol (cis- and trans- mixture)
Formel:
C₉H₁₈O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00091618 CAS-Nummer: 52204-65-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...