1-Brom-4-octylbenzol

Lieferant: Apollo Scientific
Achtung

Synonyme: p-Brom-octylbenzol

OR21923-1G
APOSOR21923-1GEA 21.9 EUR
APOSOR21923-1G
1-Brom-4-octylbenzol
1-Brom-4-octylbenzol
Formel: C₁₄H₂₁Br
Molecular Weight: 269,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00038348
CAS-Nummer: 51554-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR