1-Brom-4-hexylbenzol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
p-Brom-hexylbenzol
OR21925-100G
OR21925-25G
APOSOR21925-100GEA
313
EUR
APOSOR21925-100G
APOSOR21925-25G
1-Brom-4-hexylbenzol 97%
1-Brom-4-hexylbenzol
Formel:
C₁₂H₁₇Br Molecular Weight: 241.17 g/mol Siedepunkt: 145 °C/0.75mmHg Dichte: 1.18 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061114 CAS-Nummer: 23703-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...