3,5-Di-tert-butyl-4-hydroxyphenylacetonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR21939-1G
APOSOR21939-1GEA
174
EUR
APOSOR21939-1G
3,5-Di-tert-butyl-4-hydroxyphenylacetonitrile
3,5-Di-tert-butyl-4-hydroxyphenylacetonitrile
Formel:
C₁₆H₂₃NO Schmelzpunkt: 104…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156139 CAS-Nummer: 1611-07-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...