Methyl-2-brom-5-chlorbenzoat 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR2198-100G
OR2198-25G
APOSOR2198-100GEA
114
EUR
APOSOR2198-100G
APOSOR2198-25G
Methyl-2-brom-5-chlorbenzoat 98%
Methyl-2-brom-5-chlorbenzoat
Formel:
C₈H₆BrClO₂ Schmelzpunkt: 37…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144763 CAS-Nummer: 27007-53-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...