5-Hydroxy-4H-chromen-4-one 95%

Lieferant: Apollo Scientific

OR21982-1G
APOSOR21982-1GEA 400 EUR
APOSOR21982-1G
5-Hydroxy-4H-chromen-4-one 95%
5-Hydroxy-4H-chromen-4-one
Formel: C₉H₆O₃
Molecular Weight: 162,142 g/mol
Schmelzpunkt: 127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00100605
CAS-Nummer: 3952-69-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR